Heptafluorobutyrylamidine

Supplier: Apollo Scientific
Warning

PC4125-10G PC4125-1G PC4125-5G
APOSPC4125-10GEA 236 EUR
APOSPC4125-10G APOSPC4125-1G APOSPC4125-5G
Heptafluorobutyrylamidine
Heptafluorobutyrylamidine
Formula: C₄H₃F₇N₂
MW: 212.07 g/mol
Melting Pt: 53…55 °C
Storage Temperature: Ambient
MDL Number: MFCD00236736
CAS Number: 375-19-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR