3'-Fluoro-2-(3-fluorophenyl)acetophenone

Supplier: Apollo Scientific
Warning

PC421007-1G PC421007-5G PC421007-250MG
APOSPC421007-1GEA 139 EUR
APOSPC421007-1G APOSPC421007-5G APOSPC421007-250MG
3'-Fluoro-2-(3-fluorophenyl)acetophenone
3'-Fluoro-2-(3-fluorophenyl)acetophenone
Formula: C₁₄H₁₀F₂O
Melting Pt: 49…51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12605191
CAS Number: 40281-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR