(R)-2,2,2-Trifluoro-N-(1-phenylethyl)acetamide 95%

Supplier: Apollo Scientific

PC421193-1G
APOSPC421193-1GEA 314 EUR
APOSPC421193-1G
(R)-2,2,2-Trifluoro-N-(1-phenylethyl)acetamide 95%
(R)-2,2,2-Trifluoro-N-(1-phenylethyl)acetamide
Formula: C₁₀H₁₀F₃NO
Storage Temperature: Ambient
MDL Number: MFCD00192525
CAS Number: 39995-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR