4-Chloro-2-(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine ≥98%

Supplier: Apollo Scientific

PC421215-100MG
APOSPC421215-100MGEA 161 EUR
APOSPC421215-100MG
4-Chloro-2-(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine ≥98%
4-Chloro-2-(trifluoromethyl)-7H-pyrrolo[2,3-d]pyrimidine
Formula: C₇H₃ClF₃N₃
Storage Temperature: Ambient
MDL Number: MFCD13189354
CAS Number: 1060815-85-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR