1-Piperidinecarboxylic acid-3-fluoro-4-[(phenylmethyl)amino]-(3S,4R)-1,1-dimethylethyl ester

Supplier: Apollo Scientific

Synonyms: tert-Butyl 3,4-cis-4-(benzylamino)-3-fluoropiperidine-1-carboxylate racemate

PC430149-1G
APOSPC430149-1GEA 865 EUR
APOSPC430149-1G
1-Piperidinecarboxylic acid-3-fluoro-4-[(phenylmethyl)amino]-(3S,4R)-1,1-dimethylethyl ester
1-Piperidinecarboxylic acid-3-fluoro-4-[(phenylmethyl)amino]-(3S,4R)-1,1-dimethylethyl ester
Formula: C₁₇H₂₅FN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD22421729
CAS Number: 907572-24-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR