1-(Piperidin-4-yl)-1H-indole trifluoroacetate

Supplier: Apollo Scientific

PC430330-1G
APOSPC430330-1GEA 865 EUR
APOSPC430330-1G
1-(Piperidin-4-yl)-1H-indole trifluoroacetate
1-(Piperidin-4-yl)-1H-indole trifluoroacetate
Formula: C₁₃H₁₆N₂·CF₃CO₂H
MW: 314.303 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12912602
CAS Number: 1198286-07-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR