3-Fluoro-4-nitrobenzoic acid 97%

Supplier: Apollo Scientific
Warning

PC4399-100G PC4399-25G PC4399-5G
APOSPC4399-100GEA 116 EUR
APOSPC4399-100G APOSPC4399-25G APOSPC4399-5G
3-Fluoro-4-nitrobenzoic acid 97%
3-Fluoro-4-nitrobenzoic acid
Formula: C₇H₄FNO₄
Melting Pt: 173…176 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01862092
CAS Number: 403-21-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR