1H,1H-Heptafluoro-n-butyl methacrylate 97%

Supplier: Apollo Scientific
Warning

PC4412-25G PC4412-5G PC4412-100G
APOSPC4412-25GEA 224 EUR
APOSPC4412-25G APOSPC4412-5G APOSPC4412-100G
1H,1H-Heptafluoro-n-butyl methacrylate 97%
1H,1H-Heptafluoro-n-butyl methacrylate
Formula: C₈H₇F₇O₂
Boiling Pt: 134…136 °C
Density: 1,345 g/cm³ (25 °C)
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00039251
CAS Number: 13695-31-3
EINECS: 237-217-7
UN: 3272
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR