2-Chloro-5-trifluoromethyl-pyridine-3-sulphonyl chloride 95%

Supplier: Apollo Scientific
Danger

PC446053-500MG
APOSPC446053-500MGEA 318 EUR
APOSPC446053-500MG
2-Chloro-5-trifluoromethyl-pyridine-3-sulphonyl chloride 95%
2-Chloro-5-trifluoromethyl-pyridine-3-sulphonyl chloride
Formula: C₆H₂Cl₂F₃NO₂S
Storage Temperature: Ambient
MDL Number: MFCD15142817
CAS Number: 1208081-79-1
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR