1-(3-(Trifluoromethyl)-2-pyridinyl)piperazine hydrochloride 95+%

Supplier: Apollo Scientific

PC446085-1G
APOSPC446085-1GEA 91.5 EUR
APOSPC446085-1G
1-(3-(Trifluoromethyl)-2-pyridinyl)piperazine hydrochloride 95+%
1-(3-(Trifluoromethyl)-2-pyridinyl)piperazine hydrochloride
Formula: C₁₀H₁₂F₃N₃.HCl
Storage Temperature: Ambient
MDL Number: MFCD00114705
CAS Number: 87394-50-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR