2-(4-Trifluoromethylphenyl)piperazine

Supplier: Apollo Scientific

PC449016-1G
APOSPC449016-1GEA 112 EUR
APOSPC449016-1G
2-(4-Trifluoromethylphenyl)piperazine
2-(4-Trifluoromethylphenyl)piperazine
Formula: C₁₁H₁₃F₃N₂
Storage Temperature: Ambient
MDL Number: MFCD05864678
CAS Number: 185110-19-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR