4-(2-Fluoroethyl)piperidine hydrochloride

Supplier: Apollo Scientific
Warning

PC450041-100MG PC450041-250MG
APOSPC450041-100MGEA 116 EUR
APOSPC450041-100MG APOSPC450041-250MG
4-(2-Fluoroethyl)piperidine hydrochloride
4-(2-Fluoroethyl)piperidine hydrochloride
Formula: C₇H₁₄FN·ClH
Storage Temperature: Ambient
MDL Number: MFCD27918502
CAS Number: 184044-25-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR