4-Chlorophenyl trifluoromethanesulphonate 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-Chlorophenyltriflate, 4-Chlorophenyl triflate

PC4504-1G PC4504-25G PC4504-5G
APOSPC4504-1GEA 21.9 EUR
APOSPC4504-1G APOSPC4504-25G APOSPC4504-5G
4-Chlorophenyl trifluoromethanesulphonate 97%
4-Chlorophenyl trifluoromethanesulphonate
Formula: C₇H₄ClF₃O₃S
MW: 260.618 g/mol
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07784323
CAS Number: 29540-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR