Butyl 2,2,3,3,3-pentafluoropropyl carbonate

Supplier: Apollo Scientific

PC450446-25G
APOSPC450446-25GEA 1540 EUR
APOSPC450446-25G
Butyl 2,2,3,3,3-pentafluoropropyl carbonate
Butyl 2,2,3,3,3-pentafluoropropyl carbonate
Formula: C₈H₁₁F₅O₃
Storage Temperature: Ambient
MDL Number: MFCD28334420
CAS Number: 1923267-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR