sec-Butyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate

Supplier: Apollo Scientific

PC450475-25G
APOSPC450475-25GEA 1540 EUR
APOSPC450475-25G
sec-Butyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
sec-Butyl 2,2,3,3,4,4,4-heptafluorobutyl carbonate
Formula: C₉H₁₁F₇O₃
MW: 300,171 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28334447
CAS Number: 1980038-46-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR