1-Fluoroheptane 99%

Supplier: Apollo Scientific
Danger

Synonyms: Heptyl fluoride

PC4522-1G
APOSPC4522-1GEA 62.5 EUR
APOSPC4522-1G
1-Fluoroheptane 99%
1-Fluoroheptane
Formula: C₇H₁₅F
Boiling Pt: 118…119 °C/750mmHg
Melting Pt: –73 °C
Density: 0,806 g/cm³ (25 °C)
Flash Pt: 17 °C
Storage Temperature: Ambient
CAS Number: 661-11-0
EINECS: 211-545-0
UN: 1993
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR