1-Chloro-2-(trifluoromethoxy)benzene

Supplier: Apollo Scientific
Warning

PC4580-100G PC4580-25G PC4580-500G
APOSPC4580-100GEA 58 EUR
APOSPC4580-100G APOSPC4580-25G APOSPC4580-500G
1-Chloro-2-(trifluoromethoxy)benzene
1-Chloro-2-(trifluoromethoxy)benzene
Formula: C₇H₄ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD03840206
CAS Number: 450-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR