1-(2-Chloroethyl)-3-(3-fluorophenyl)urea

Supplier: Apollo Scientific
Warning

PC46359-1G PC46359-250MG
APOSPC46359-1GEA 26.1 EUR
APOSPC46359-1G APOSPC46359-250MG
1-(2-Chloroethyl)-3-(3-fluorophenyl)urea
1-(2-Chloroethyl)-3-(3-fluorophenyl)urea
Formula: C₉H₁₀ClFN₂O
Storage Temperature: Ambient
MDL Number: MFCD14586691
CAS Number: 146257-15-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR