1-Bromo-7-fluoroisoquinoline

Supplier: Apollo Scientific
Warning

PC48317-500MG PC48317-100MG PC48317-250MG
APOSPC48317-500MGEA 220 EUR
APOSPC48317-500MG APOSPC48317-100MG APOSPC48317-250MG
1-Bromo-7-fluoroisoquinoline
1-Bromo-7-fluoroisoquinoline
Formula: C₉H₅BrFN
Storage Temperature: Refrigerator
MDL Number: MFCD15832812
CAS Number: 1207448-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR