2-Methoxy-3-(trifluoromethyl)benzonitrile 97%

Supplier: Apollo Scientific
Danger

PC48356-1G PC48356-5G PC48356-25G PC48356-100G
APOSPC48356-1GEA 63.8 EUR
APOSPC48356-1G APOSPC48356-5G APOSPC48356-25G APOSPC48356-100G
2-Methoxy-3-(trifluoromethyl)benzonitrile 97%
2-Methoxy-3-(trifluoromethyl)benzonitrile
Formula: C₉H₆F₃NO
Storage Temperature: Ambient
MDL Number: MFCD09832310
CAS Number: 1017778-62-9
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR