2-(Difluoromethyl)benzyl bromide

Supplier: Apollo Scientific
Danger

PC48365-1G PC48365-250MG
APOSPC48365-1GEA 549 EUR
APOSPC48365-1G APOSPC48365-250MG
2-(Difluoromethyl)benzyl bromide
2-(Difluoromethyl)benzyl bromide
Formula: C₈H₇BrF₂
Storage Temperature: Refrigerator
MDL Number: MFCD18791598
CAS Number: 1222556-61-7
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR