3-Hydroxy-5-(trifluoromethyl)benzonitrile

Supplier: Apollo Scientific
Danger

PC48422-1G PC48422-250MG PC48422-5G
APOSPC48422-1GEA 509 EUR
APOSPC48422-1G APOSPC48422-250MG APOSPC48422-5G
3-Hydroxy-5-(trifluoromethyl)benzonitrile
3-Hydroxy-5-(trifluoromethyl)benzonitrile
Formula: C₈H₄F₃NO
Melting Pt: 128…130 °C
Storage Temperature: Refrigerator
MDL Number: MFCD16999150
CAS Number: 1243459-56-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR