4-Bromo-2-nitro-6-(trifluoromethyl)aniline

Supplier: Apollo Scientific
Warning

PC48440-1G PC48440-5G
APOSPC48440-1GEA 52.2 EUR
APOSPC48440-1G APOSPC48440-5G
4-Bromo-2-nitro-6-(trifluoromethyl)aniline
4-Bromo-2-nitro-6-(trifluoromethyl)aniline
Formula: C₇H₄BrF₃N₂O₂
Melting Pt: 80…82 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD09835318
CAS Number: 157026-18-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR