3,5-Dichloro-2-fluorobenzoic acid

Supplier: Apollo Scientific
Warning

PC48486-1G PC48486-5G
APOSPC48486-1GEA 487 EUR
APOSPC48486-1G APOSPC48486-5G
3,5-Dichloro-2-fluorobenzoic acid
3,5-Dichloro-2-fluorobenzoic acid
Formula: C₇H₃Cl₂FO₂
Storage Temperature: Ambient
MDL Number: MFCD04091131
CAS Number: 665022-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR