3-Bromo-4-iodobenzotrifluoride

Supplier: Apollo Scientific
Warning

PC48522-1G PC48522-25G PC48522-5G
APOSPC48522-1GEA 21.9 EUR
APOSPC48522-1G APOSPC48522-25G APOSPC48522-5G
3-Bromo-4-iodobenzotrifluoride
3-Bromo-4-iodobenzotrifluoride
Formula: C₇H₃BrF₃I
Storage Temperature: Refrigerator
MDL Number: MFCD08543938
CAS Number: 481075-58-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR