3,4-Difluoro-2-methylbenzonitrile 98%

Supplier: Apollo Scientific
Danger

PC48776-5G
APOSPC48776-5GEA 178 EUR
APOSPC48776-5G
3,4-Difluoro-2-methylbenzonitrile 98%
3,4-Difluoro-2-methylbenzonitrile
Formula: C₈H₅F₂N
Melting Pt: 39…44 °C
Storage Temperature: Ambient
MDL Number: MFCD07777154
CAS Number: 847502-83-4
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR