2,3,4-Trifluoro-6-(trifluoromethyl)benzyl alcohol 95%

Supplier: Apollo Scientific
Warning

PC48777-1G PC48777-250MG PC48777-5G
APOSPC48777-1GEA 493 EUR
APOSPC48777-1G APOSPC48777-250MG APOSPC48777-5G
2,3,4-Trifluoro-6-(trifluoromethyl)benzyl alcohol 95%
2,3,4-Trifluoro-6-(trifluoromethyl)benzyl alcohol
Formula: C₈H₄F₆O
Storage Temperature: Ambient
MDL Number: MFCD31629891
CAS Number: 2244085-32-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR