6-Fluoroquinoline-3-carboxylic acid

Supplier: Apollo Scientific
Warning

PC49031-1G
APOSPC49031-1GEA 255 EUR
APOSPC49031-1G
6-Fluoroquinoline-3-carboxylic acid
6-Fluoroquinoline-3-carboxylic acid
Formula: C₁₀H₆FNO₂
Melting Pt: 269…270 °C
Storage Temperature: Ambient
MDL Number: MFCD08437578
CAS Number: 116293-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR