3-Amino-6-bromo-2-hydroxybenzotrifluoride 96%

Supplier: Apollo Scientific
Danger

PC49077-1G PC49077-250MG
APOSPC49077-1GEA 406 EUR
APOSPC49077-1G APOSPC49077-250MG
3-Amino-6-bromo-2-hydroxybenzotrifluoride 96%
3-Amino-6-bromo-2-hydroxybenzotrifluoride
Formula: C₇H₅BrF₃NO
Storage Temperature: Refrigerator
MDL Number: MFCD25541884
CAS Number: 1445995-79-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR