4'-Bromo-2,2,2,2'-tetrafluoroacetophenone

Supplier: Apollo Scientific
Warning

PC49089-1G PC49089-250MG PC49089-5G
APOSPC49089-1GEA 150 EUR
APOSPC49089-1G APOSPC49089-250MG APOSPC49089-5G
4'-Bromo-2,2,2,2'-tetrafluoroacetophenone
4'-Bromo-2,2,2,2'-tetrafluoroacetophenone
Formula: C₈H₃BrF₄O
Storage Temperature: Ambient
MDL Number: MFCD19439730
CAS Number: 617706-18-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR