2-Fluoro-4-iodonitrobenzene

Supplier: Apollo Scientific
Warning

PC49098-1G PC49098-5G PC49098-25G PC49098-10G
APOSPC49098-1GEA 40.6 EUR
APOSPC49098-1G APOSPC49098-5G APOSPC49098-25G APOSPC49098-10G
2-Fluoro-4-iodonitrobenzene
2-Fluoro-4-iodonitrobenzene
Formula: C₆H₃FINO₂
Melting Pt: 118…118,5 °C
Storage Temperature: Ambient
MDL Number: MFCD18396989
CAS Number: 2996-31-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR