5-Chloro-2,4-difluorobenzonitrile 97%

Supplier: Apollo Scientific
Warning

PC49273-1G PC49273-5G PC49273-10G
APOSPC49273-1GEA 24.8 EUR
APOSPC49273-1G APOSPC49273-5G APOSPC49273-10G
5-Chloro-2,4-difluorobenzonitrile 97%
5-Chloro-2,4-difluorobenzonitrile
Formula: C₇H₂ClF₂N
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD06659465
CAS Number: 146780-26-9
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR