4-(Hydroxymethyl)-2-(trifluoromethyl)pyridine 95%

Supplier: Apollo Scientific
Warning

PC49314-1G PC49314-250MG PC49314-500MG PC49314-5G
APOSPC49314-1GEA 311 EUR
APOSPC49314-1G APOSPC49314-250MG APOSPC49314-500MG APOSPC49314-5G
4-(Hydroxymethyl)-2-(trifluoromethyl)pyridine 95%
4-(Hydroxymethyl)-2-(trifluoromethyl)pyridine
Formula: C₇H₆F₃NO
Storage Temperature: Ambient
MDL Number: MFCD11100737
CAS Number: 131747-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR