5-Bromo-2,3,4-trifluorobenzyl bromide 95%

Supplier: Apollo Scientific
Danger

PC49489-1G PC49489-250MG PC49489-5G
APOSPC49489-1GEA 58 EUR
APOSPC49489-1G APOSPC49489-250MG APOSPC49489-5G
5-Bromo-2,3,4-trifluorobenzyl bromide 95%
5-Bromo-2,3,4-trifluorobenzyl bromide
Formula: C₇H₃Br₂F₃
Storage Temperature: Ambient
MDL Number: MFCD12756772
CAS Number: 1260832-85-6
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR