Iron(II) tetrafluoroborate hexahydrate 97%

Supplier: Apollo Scientific

Synonyms: Ferrous tetrafluoroborate

PC49590-100G PC49590-25G
APOSPC49590-100GEA 215 EUR
APOSPC49590-100G APOSPC49590-25G
Iron(II) tetrafluoroborate hexahydrate 97%
Iron(II) tetrafluoroborate hexahydrate
Formula: 2(BF₄)·Fe·6H₂O
Storage Temperature: Ambient
MDL Number: MFCD00192235
CAS Number: 13877-16-2
UN: 3260
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR