4-Bromo-3,5-difluorobenzenesulphonyl chloride 97%

Supplier: Apollo Scientific
Danger

PC4965-1G PC4965-5G PC4965-25G
APOSPC4965-1GEA 21.9 EUR
APOSPC4965-1G APOSPC4965-5G APOSPC4965-25G
4-Bromo-3,5-difluorobenzenesulphonyl chloride 97%
4-Bromo-3,5-difluorobenzenesulphonyl chloride
Formula: C₆H₂BrClF₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD03788548
CAS Number: 518057-63-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR