5-Bromo-2-(trifluoromethoxy)phenol 98%

Supplier: Apollo Scientific

PC499007-1G
APOSPC499007-1GEA 139 EUR
APOSPC499007-1G
5-Bromo-2-(trifluoromethoxy)phenol 98%
5-Bromo-2-(trifluoromethoxy)phenol
Formula: C₇H₄BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD21333031
CAS Number: 1048963-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR