2,3-Difluoro-4-(trifluoromethyl)benzonitrile 98%

Supplier: Apollo Scientific
Danger

PC4993-1G PC4993-250MG PC4993-5G
APOSPC4993-1GEA 258 EUR
APOSPC4993-1G APOSPC4993-250MG APOSPC4993-5G
2,3-Difluoro-4-(trifluoromethyl)benzonitrile 98%
2,3-Difluoro-4-(trifluoromethyl)benzonitrile
Formula: C₈H₂F₅N
Storage Temperature: Ambient
MDL Number: MFCD00236258
CAS Number: 240122-24-1
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR