Methyl 2-bromo-6-chloro-4-fluorobenzoate 95%

Supplier: Apollo Scientific
Warning

PC49997-1G PC49997-250MG
APOSPC49997-1GEA 219 EUR
APOSPC49997-1G APOSPC49997-250MG
Methyl 2-bromo-6-chloro-4-fluorobenzoate 95%
Methyl 2-bromo-6-chloro-4-fluorobenzoate
Formula: C₈H₅BrClFO₂
Storage Temperature: Ambient
MDL Number: MFCD28733926
CAS Number: 1695489-54-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR