3-Fluoro-5-nitrobenzyl bromide

Supplier: Apollo Scientific
Danger

PC501426-1G PC501426-5G
APOSPC501426-1GEA 406 EUR
APOSPC501426-1G APOSPC501426-5G
3-Fluoro-5-nitrobenzyl bromide
3-Fluoro-5-nitrobenzyl bromide
Formula: C₇H₅BrFNO₂
Storage Temperature: Ambient
MDL Number: MFCD13185623
CAS Number: 883987-75-5
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR