3’-Bromo-2’,4’-difluoroacetophenone 95%

Supplier: Apollo Scientific
Warning

PC501684-1G PC501684-5G PC501684-25G
APOSPC501684-1GEA 21.9 EUR
APOSPC501684-1G APOSPC501684-5G APOSPC501684-25G
3’-Bromo-2’,4’-difluoroacetophenone 95%
3’-Bromo-2’,4’-difluoroacetophenone
Formula: C₈H₅BrF₂O
Storage Temperature: Ambient
MDL Number: MFCD13689251
CAS Number: 1210824-63-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR