4-Fluoro-5-methoxybenzene-1,2-diamine

Supplier: Apollo Scientific

PC501750-1G
APOSPC501750-1GEA 611 EUR
APOSPC501750-1G
4-Fluoro-5-methoxybenzene-1,2-diamine
4-Fluoro-5-methoxybenzene-1,2-diamine
Formula: C₇H₉FN₂O
MW: 156,158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18804706
CAS Number: 125163-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR