Methyl 5-bromo-2-(trifluoromethyl)pyridine-3-carboxylate 95%

Supplier: Apollo Scientific

PC50187-1G PC50187-250MG
APOSPC50187-1GEA 0 EUR
APOSPC50187-1G APOSPC50187-250MG
Methyl 5-bromo-2-(trifluoromethyl)pyridine-3-carboxylate 95%
Methyl 5-bromo-2-(trifluoromethyl)pyridine-3-carboxylate
Formula: C₈H₅BrF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD16038782
CAS Number: 850864-56-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR