3-Bromo-2-(trifluoromethyl)benzyl alcohol

Supplier: Apollo Scientific
Warning

PC502065-1G PC502065-250MG PC502065-5G
APOSPC502065-1GEA 226 EUR
APOSPC502065-1G APOSPC502065-250MG APOSPC502065-5G
3-Bromo-2-(trifluoromethyl)benzyl alcohol
3-Bromo-2-(trifluoromethyl)benzyl alcohol
Formula: C₈H₆BrF₃O
Storage Temperature: Ambient
MDL Number: MFCD28746751
CAS Number: 1805950-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR