2-Chloro-6-(trifluoromethoxy)benzyl bromide 95%

Supplier: Apollo Scientific
Danger

PC502717-1G PC502717-250MG
APOSPC502717-1GEA 609 EUR
APOSPC502717-1G APOSPC502717-250MG
2-Chloro-6-(trifluoromethoxy)benzyl bromide 95%
2-Chloro-6-(trifluoromethoxy)benzyl bromide
Formula: C₈H₅BrClF₃O
Storage Temperature: Ambient
MDL Number: MFCD18393831
CAS Number: 1261822-79-0
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR