3-Bromo-4-(trifluoromethyl)benzyl fluoride 95%

Supplier: Apollo Scientific
Danger

PC502899-1G PC502899-250MG
APOSPC502899-1GEA 609 EUR
APOSPC502899-1G APOSPC502899-250MG
3-Bromo-4-(trifluoromethyl)benzyl fluoride 95%
3-Bromo-4-(trifluoromethyl)benzyl fluoride
Formula: C₈H₅BrF₄
Storage Temperature: Ambient
MDL Number: MFCD31634835
CAS Number: 2244087-06-5
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR