2-Chloro-3-fluoro-hydrazine hydrochloride 98%

Supplier: Apollo Scientific
Warning

Synonyms: (2-Chloro-3-fluorophenyl)hydrazine hydrochloride

PC50314-25G
APOSPC50314-25GEA 186 EUR
APOSPC50314-25G
2-Chloro-3-fluoro-hydrazine hydrochloride 98%
2-Chloro-3-fluoro-hydrazine hydrochloride
Formula: C₆H₆ClFN₂·ClH
MW: 197.038 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18393091
CAS Number: 1138036-54-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR