N-(2,2,2-Trifluoroethyl)-4-bromobenzylamine 97%

Supplier: Apollo Scientific
Warning

PC50332-1G PC50332-250MG PC50332-5G
APOSPC50332-1GEA 480 EUR
APOSPC50332-1G APOSPC50332-250MG APOSPC50332-5G
N-(2,2,2-Trifluoroethyl)-4-bromobenzylamine 97%
N-(2,2,2-Trifluoroethyl)-4-bromobenzylamine
Formula: C₉H₉BrF₃N
MW: 268.074 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09928975
CAS Number: 728948-25-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR