2-Ethynyl-3-fluorobenzonitrile

Supplier: Apollo Scientific

PC50557-5G
APOSPC50557-5GEA 4250 EUR
APOSPC50557-5G
2-Ethynyl-3-fluorobenzonitrile
2-Ethynyl-3-fluorobenzonitrile
Formula: C₉H₄FN
Storage Temperature: Freezer
CAS Number: 1848959-59-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR