2,4,5-Trifluoro-3-methoxybenzoic acid 98%

Supplier: Apollo Scientific
Warning

PC5060T-100G PC5060T-25G PC5060T-500G
APOSPC5060T-100GEA 43.5 EUR
APOSPC5060T-100G APOSPC5060T-25G APOSPC5060T-500G
2,4,5-Trifluoro-3-methoxybenzoic acid 98%
2,4,5-Trifluoro-3-methoxybenzoic acid
Formula: C₈H₅F₃O₃
Melting Pt: 105…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00153201
CAS Number: 112811-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR